Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-(2-Vinylanthracene-9,10-diylidene)dimalononitrile 98.0+%, TCI America™
SDP

Catalog No. V01375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01375G 5 g
V01371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01375G Supplier TCI America Supplier No. V01375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1612793-07-3
Molecular Formula C22H10N4
Molecular Weight (g/mol) 330.35
InChI Key SUTLXPZYDBOIRV-UHFFFAOYSA-N
Synonym 9,10-Bis(dicyanomethylene)-2-vinylanthracene
PubChem CID 91972128
IUPAC Name 2-[10-(dicyanomethylidene)-3-ethenylanthracen-9-ylidene]propanedinitrile
SMILES C=CC1=CC2=C(C=C1)C(=C(C#N)C#N)C3=CC=CC=C3C2=C(C#N)C#N

Specifications

Quantity 5 g
UN Number 3439
Formula Weight 330.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-(2-Vinylanthracene-9,10-diylidene)dimalononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.