accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONO

CAS RN 161282-57-1

IUPAC Name: tert-butyl 2-formyl-1H-pyrrole-1-carboxylate
Synonyms: tert-butyl 2-formylpyrrole-1-carboxylate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: NSHLHMBABZKJHP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results