CAS RN 1612886-77-7
IUPAC Name:
4-phenyl-2-(pyridin-4-yl)-1,3-thiazole-5-carbonitrile
Synonyms:
4-phenyl-2-(pyridin-4-yl)-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol):
263.32
Molecular Formula:
C15H9N3S
InChi Key:
FAOMVWUEFLWNPR-UHFFFAOYSA-N
SMILES:
N#CC1=C(N=C(S1)C1=CC=NC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Cyano-4-phenyl-2-(4-pyridyl)thiazole, 97%, Thermo Scientific™
CAS: 1612886-77-7 Molecular Formula: C15H9N3S Molecular Weight (g/mol): 263.318 MDL Number: MFCD22683013 InChI Key: FAOMVWUEFLWNPR-UHFFFAOYSA-N Synonym: 4-phenyl-2-pyridin-4-yl-1,3-thiazole-5-carbonitrile,5-cyano-4-phenyl-2-4-pyridyl thiazole PubChem CID: 90643528 IUPAC Name: 4-phenyl-2-pyridin-4-yl-1,3-thiazole-5-carbonitrile SMILES: C1=CC=C(C=C1)C2=C(SC(=N2)C3=CC=NC=C3)C#N