accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1614-82-0

IUPAC Name: 2,7-dimethoxy-1,4,5,8-tetrahydronaphthalene
Synonyms: 2,7-dimethoxy-1,4,5,8-tetrahydronaphthalene
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: PWPPYWWWEVISEX-UHFFFAOYSA-N
SMILES: COC1=CCC2=C(C1)CC(OC)=CC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results