Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalene 97.0+%, TCI America™
SDP

Catalog No. D27325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27325G Supplier TCI America Supplier No. D27325G

Chemical Identifiers

CAS 1614-82-0
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.26
MDL Number MFCD00269695
InChI Key PWPPYWWWEVISEX-UHFFFAOYSA-N
PubChem CID 13950331
IUPAC Name 2,7-dimethoxy-1,4,5,8-tetrahydronaphthalene
SMILES COC1=CCC2=C(C1)CC(OC)=CC2

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.