CAS RN 161401-25-8
CAS RN 161401-25-8
IUPAC Name:
tetramethylazanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms:
bistriflylimide anion; tetramethylammonium ion
Molecular Weight (g/mol):
354.28
Molecular Formula:
C6H12F6N2O4S2
InChi Key:
JTCSZQBQVDPVMT-UHFFFAOYSA-N
SMILES:
C[N+](C)(C)C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich Fmoc-Lys(Palmitoyl)-Oh
Sigma Aldrich Fmoc-Lys(Palmitoyl)-Oh
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.