CAS RN 161401-82-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Caspase Inhibitor VI
Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.