Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Inhibitor VI

Catalog No. 219007250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-900-7250UG 250 μg
21-900-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-900-7250UG Supplier MilliporeSigma™ Supplier No. 219007250UG
Add to Cart
Add to Cart

Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.

Peptide Sequence: Z-Val-Ala-Asp-CH2F*
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins general caspase inhibitor
CAS 161401-82-7
Molecular Formula C21H28FN3O7
Formula Weight 453.5g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 250 μg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.