accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16144-91-5

IUPAC Name: (1R,8S)-tricyclo[6.2.1.0²,⁷]undeca-2(7),3,5-triene-3,6-diol
Synonyms: (1R,8S)-tricyclo[6.2.1.0²,⁷]undeca-2(7),3,5-triene-3,6-diol
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: JYHNNCBQCSLFQM-KNVOCYPGSA-N
SMILES: OC1=CC=C(O)C2=C1[C@H]1CC[C@@H]2C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)