Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dihydroxybenzonorbornane 98.0+%, TCI America™
SDP

Catalog No. D18065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18065G 5 g
D180625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18065G Supplier TCI America Supplier No. D18065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16144-91-5
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00077441
InChI Key JYHNNCBQCSLFQM-UHFFFAOYSA-N
Synonym Benzonorbornene-3,6-diol, 1,2,3,4-Tetrahydro-1,4-methanonaphthalene-5,8-diol
PubChem CID 85967
SMILES C1CC2CC1C3=C(C=CC(=C23)O)O

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 176.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dihydroxybenzonorbornane [Antioxidant]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.