accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNSOHO

CAS RN 161512-71-6

IUPAC Name: 3-(benzylsulfanyl)-2-acetamidopropanoic acid
Synonyms: 3-(benzylsulfanyl)-2-acetamidopropanoic acid
Molecular Weight (g/mol): 253.32
Molecular Formula: C12H15NO3S
InChi Key: BJUXDERNWYKSIQ-UHFFFAOYNA-N
SMILES: CC(=O)NC(CSCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results