Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-S-benzyl-D-cysteine, TCI America™
SDP

Catalog No. A0740100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0740100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0740100MG Supplier TCI America Supplier No. A0740100MG

Chemical Identifiers

CAS 161512-71-6
Molecular Formula C12H15NO3S
Molecular Weight (g/mol) 253.316
MDL Number MFCD00070565
InChI Key BJUXDERNWYKSIQ-LLVKDONJSA-N
Synonym D-Benzylmercapturic Acid, Ac-D-Cys(Bzl)-OH
PubChem CID 853882
IUPAC Name (2S)-2-acetamido-3-benzylsulfanylpropanoic acid
SMILES CC(=O)NC(CSCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 100 mg
Formula Weight 253.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-S-benzyl-D-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.