accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3BOHHO

CAS RN 16152-51-5

IUPAC Name: [4-(propan-2-yl)phenyl]boronic acid
Synonyms: 4-isopropylphenylboronic acid
Molecular Weight (g/mol): 164.01
Molecular Formula: C9H13BO2
InChi Key: IAEUFBDMVKQCLU-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results