Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-isopropylphenylboronic acid

Catalog No. CDS013712 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013712 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013712 Supplier Sigma Aldrich Supplier No. CDS013712500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(isopropylamino)ethanol is a secondary amine
  • The solubility of co2 in aqueous solutions of 2-(isopropylamino)ethanol (ipae) has been evaluated.2-(isopropylamino)ethanol may be used in the synthesis of heterocyclic n,o-acetals

Specifications

CAS 16152-51-5
Molecular Formula C9H13BO2
Linear Formula C9H13BO2
MDL Number MFCD01074614
Quantity 250 mL
Molecular Weight (g/mol) 164.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.