CAS RN 16156-50-6
IUPAC Name:
hexyl methanesulfonate
Synonyms:
hexyl methanesulfonate
Molecular Weight (g/mol):
180.26
Molecular Formula:
C7H16O3S
InChi Key:
URIRDRHUUFRHAS-UHFFFAOYSA-N
SMILES:
CCCCCCOS(C)(=O)=O
1
–
1
of
1
results
1
Hexyl Methanesulfonate 96.0+%, TCI America™
CAS: 16156-50-6 Molecular Formula: C7H16O3S Molecular Weight (g/mol): 180.26 MDL Number: MFCD00674707 InChI Key: URIRDRHUUFRHAS-UHFFFAOYSA-N Synonym: Methanesulfonic Acid Hexyl Ester PubChem CID: 12517373 IUPAC Name: hexyl methanesulfonate SMILES: CCCCCCOS(C)(=O)=O