accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OSCH3OO

CAS RN 16156-53-9

IUPAC Name: 2-methylpropyl methanesulfonate
Synonyms: 2-methylpropyl methanesulfonate
Molecular Weight (g/mol): 152.21
Molecular Formula: C5H12O3S
InChi Key: PZMKMCFRTKGVOL-UHFFFAOYSA-N
SMILES: CC(C)COS(C)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results