CAS RN 16159-70-9
CAS RN 16159-70-9
IUPAC Name:
4-nitrophenyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Synonyms:
4-nitrophenyl 2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoate
Molecular Weight (g/mol):
386.4
Molecular Formula:
C20H22N2O6
InChi Key:
QZIWWFMMLBBICG-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Phe-ONp, Advanced ChemTech
CAS: 16159-70-9 Molecular Formula: C20H22N2O6 Molecular Weight (g/mol): 386.40 InChI Key: QZIWWFMMLBBICG-UHFFFAOYNA-N IUPAC Name: 4-nitrophenyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O