accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOOHCNH(R)

CAS RN 161735-79-1

IUPAC Name: (1R)-N-(prop-2-yn-1-yl)-2,3-dihydro-1H-inden-1-amine; methanesulfonic acid
Synonyms: methanesulfonic acid; rasagiline
Molecular Weight (g/mol): 267.34
Molecular Formula: C13H17NO3S
InChi Key: JDBJJCWRXSVHOQ-UTONKHPSSA-N
SMILES: CS(O)(=O)=O.C#CCN[C@@H]1CCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results