accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOOCl

CAS RN 16191-84-7

IUPAC Name: 1-chloro-4-(2-chloroethanesulfonyl)benzene
Synonyms: 1-chloro-4-(2-chloroethanesulfonyl)benzene
Molecular Weight (g/mol): 239.11
Molecular Formula: C8H8Cl2O2S
InChi Key: KXQHTLXSDBXWNB-UHFFFAOYSA-N
SMILES: ClCCS(=O)(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results