accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHH3COOCH3CH3

CAS RN 16200-50-3

IUPAC Name: ethyl 3,4-diethyl-5-methyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 3,4-diethyl-5-methyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 209.29
Molecular Formula: C12H19NO2
InChi Key: YXAABSBFWADBRO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(CC)C(CC)=C(C)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results