Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole 96.0+%, TCI America™
SDP

Catalog No. D22175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22175G 5 g
D22171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22175G Supplier TCI America Supplier No. D22175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16200-50-3
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.289
MDL Number MFCD00075089
InChI Key YXAABSBFWADBRO-UHFFFAOYSA-N
Synonym 3,4-Diethyl-5-methyl-2-pyrrolecarboxylic Acid Ethyl Ester, Ethyl 3,4-Diethyl-5-methyl-2-pyrrolecarboxylate
PubChem CID 590710
IUPAC Name ethyl 3,4-diethyl-5-methyl-1H-pyrrole-2-carboxylate
SMILES CCC1=C(NC(=C1CC)C(=O)OCC)C

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
Formula Weight 209.29
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.