CAS RN 162011-90-7
IUPAC Name:
4-(4-methanesulfonylphenyl)-3-phenyl-2,5-dihydrofuran-2-one
Synonyms:
rofecoxib vioxx ceoxx
Molecular Weight (g/mol):
314.36
Molecular Formula:
C17H14O4S
InChi Key:
RZJQGNCSTQAWON-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=C(C=C1)C1=C(C(=O)OC1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Rofecoxib
Thermo Scientific Chemicals Rofecoxib
CAS: 162011-90-7 Molecular Formula: C17 H14 O4 S