accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOFHNNO

CAS RN 162012-69-3

IUPAC Name: 7-fluoro-6-nitro-1,4-dihydroquinazolin-4-one
Synonyms: 7-fluoro-6-nitro-1H-quinazolin-4-one
Molecular Weight (g/mol): 209.14
Molecular Formula: C8H4FN3O3
InChi Key: VTUAEMSZEIGQRM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C2NC=NC(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results