Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluoro-6-nitroquinazolin-4(1H)-one 98.0+%, TCI America™
SDP

Catalog No. F10911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10911G Supplier TCI America Supplier No. F10911G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 162012-69-3
Molecular Formula C8H4FN3O3
Molecular Weight (g/mol) 209.136
MDL Number MFCD08690890
InChI Key VTUAEMSZEIGQRM-UHFFFAOYSA-N
Synonym 7-Fluoro-6-nitroquinazolin-4(3H)-one, 7-Fluoro-4-hydroxy-6-nitroquinazoline
PubChem CID 7063911
IUPAC Name 7-fluoro-6-nitro-1H-quinazolin-4-one
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])F)NC=NC2=O

Specifications

Melting Point 288°C
Quantity 1 g
Formula Weight 209.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Fluoro-6-nitroquinazolin-4(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.