accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHONHNO(E/Z)(E/Z)

CAS RN 1620285-48-4

IUPAC Name: 3,3'-bis({[2-(2,3-dihydro-1H-isoindol-2-yl)-1,2-diphenylethyl]amino}methylidene)-2H,2'H,3H,3'H-[1,1'-binaphthalene]-2,2'-dione
Synonyms: 3,3'-bis({[2-(1,3-dihydroisoindol-2-yl)-1,2-diphenylethyl]amino}methylidene)-[1,1'-binaphthalene]-2,2'-dione
Molecular Weight (g/mol): 935.18
Molecular Formula: C66H54N4O2
InChi Key: SYUJAGVYZYZTBH-UHFFFAOYNA-N
SMILES: O=C1C(=CNC(C(N2CC3=CC=CC=C3C2)C2=CC=CC=C2)C2=CC=CC=C2)C=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC(=CNC(C(N2CC3=CC=CC=C3C2)C2=CC=CC=C2)C2=CC=CC=C2)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results