Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3,3'-Bis[[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]-1,1'-bi-2-naphthol 92.0+%, TCI America™
SDP

Catalog No. B448550MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B448550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B448550MG Supplier TCI America Supplier No. B448550MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1620285-48-4
Molecular Formula C66H54N4O2
Molecular Weight (g/mol) 935.184
InChI Key SYUJAGVYZYZTBH-HODAIVDYSA-N
PubChem CID 131674832
IUPAC Name (3Z)-3-[[[(1R,2R)-2-(1,3-dihydroisoindol-2-yl)-1,2-diphenylethyl]amino]methylidene]-1-[(3Z)-3-[[[(1R,2R)-2-(1,3-dihydroisoindol-2-yl)-1,2-diphenylethyl]amino]methylidene]-2-oxonaphthalen-1-yl]naphthalen-2-one
SMILES C1C2=CC=CC=C2CN1C(C3=CC=CC=C3)C(C4=CC=CC=C4)NC=C5C=C6C=CC=CC6=C(C5=O)C7=C8C=CC=CC8=CC(=CNC(C9=CC=CC=C9)C(C1=CC=CC=C1)N1CC2=CC=CC=C2C1)C7=O

Specifications

Quantity 50 mg
Formula Weight 935.18
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3,3′-Bis[[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]-1,1′-bi-2-nap
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.