accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3CH3SCH3H3CCH3PCH3CH3CH3H3CClRu(?)(?)

CAS RN 1621182-04-4

IUPAC Name: [2',4',6'-trimethyl-3-(2,4,6-trimethylphenyl)-[1,1'-biphenyl]-2-yl]sulfanide; chlororutheniumylium; tris(propan-2-yl)phosphane
Synonyms: [2',4',6'-trimethyl-3-(2,4,6-trimethylphenyl)-[1,1'-biphenyl]-2-yl]sulfanide; chlororutheniumylium; triisopropylphosphine
Molecular Weight (g/mol): 642.28
Molecular Formula: C33H46ClPRuS
InChi Key: OLQLCTCWEPROKL-UHFFFAOYSA-L
SMILES: Cl[Ru+].CC(C)P(C(C)C)C(C)C.CC1=CC(C)=C(C(C)=C1)C1=CC=CC(=C1[S-])C1=C(C)C=C(C)C=C1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results