accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNClCl

CAS RN 16234-14-3

IUPAC Name: 2,4-dichlorothieno[3,2-d]pyrimidine
Synonyms: 2,4-dichlorothieno[3,2-d]pyrimidine
Molecular Weight (g/mol): 205.06
Molecular Formula: C6H2Cl2N2S
InChi Key: AQECFYPZMBRCIA-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C2SC=CC2=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results