Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorothieno[3,2-d]pyrimidine 98.0+%, TCI America™
SDP

Catalog No. D48641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48641G Supplier TCI America Supplier No. D48641G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16234-14-3
Molecular Formula C6H2Cl2N2S
Molecular Weight (g/mol) 205.056
MDL Number MFCD08448158
InChI Key AQECFYPZMBRCIA-UHFFFAOYSA-N
Synonym 2,4-dichlorothieno 3,2-d pyrimidine, 2,4-dichlorothiopheno 3,2-d pyrimidine, thieno 3,2-d pyrimidine, 2,4-dichloro, 2,4-dichloro-thieno 3,2-d pyrimidine, pubchem14660, 2-4, dochlorothieno 3,2d pyrimidine, 2.4-dichlorothieno 3,2-d pyrimidine, 2.4-dichloro-thieno 3,2-d pyrimidine, 2,4-bis chloranyl thieno 3,2-d pyrimidine
PubChem CID 12712832
IUPAC Name 2,4-dichlorothieno[3,2-d]pyrimidine
SMILES C1=CSC2=C1N=C(N=C2Cl)Cl

Specifications

Melting Point 138°C
Color White
Quantity 1 g
Formula Weight 205.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorothieno[3,2-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.