CAS RN 1624-62-0
CAS RN 1624-62-0
IUPAC Name:
(3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
Synonyms:
(3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-2H,3H,3aH,3bH,4H,5H,9bH,10H,11H-cyclopenta[a]phenanthren-1-one
Molecular Weight (g/mol):
284.4
Molecular Formula:
C19H24O2
InChi Key:
BCWWDWHFBMPLFQ-VXNCWWDNSA-N
SMILES:
COC1=CC=C2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCC2=C1
1
–
1
of
1
results
1
Estrone 3-Methyl Ether, TCI America™
CAS: 1624-62-0 Molecular Formula: C19H24O2 Molecular Weight (g/mol): 284.399 MDL Number: MFCD00046228 InChI Key: BCWWDWHFBMPLFQ-VXNCWWDNSA-N Synonym: 3-Methoxyestrone, 3-Methoxyestra-1,3,5(10)-trien-17-one PubChem CID: 92895 IUPAC Name: (8R,9S,13S,14S)-3-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one SMILES: CC12CCC3C(C1CCC2=O)CCC4=C3C=CC(=C4)OC