Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrone 3-Methyl Ether, TCI America™
SDP

Catalog No. E09171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09171G Supplier TCI America Supplier No. E09171G

Chemical Identifiers

CAS 1624-62-0
Molecular Formula C19H24O2
Molecular Weight (g/mol) 284.399
MDL Number MFCD00046228
InChI Key BCWWDWHFBMPLFQ-VXNCWWDNSA-N
Synonym 3-Methoxyestrone, 3-Methoxyestra-1,3,5(10)-trien-17-one
PubChem CID 92895
IUPAC Name (8R,9S,13S,14S)-3-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
SMILES CC12CCC3C(C1CCC2=O)CCC4=C3C=CC(=C4)OC

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 1 g
Formula Weight 284.40
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Estrone 3-Methyl Ether
RTECSNumber KG7775000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.