accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClGeH

CAS RN 1626-24-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 341.41
Molecular Formula: C18H17ClGe
InChi Key: BYJWKGIYYBGICW-UHFFFAOYSA-N
SMILES: Cl.C1=CC=C(C=C1)[GeH](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)