Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylchlorogermane 97.0+%, TCI America™
SDP

Catalog No. T11621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11621G Supplier TCI America Supplier No. T11621G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1626-24-0
Molecular Formula C18H15ClGe
Molecular Weight (g/mol) 339.398
MDL Number MFCD00000461
InChI Key ONQMBYVVEVFYRP-UHFFFAOYSA-N
Synonym Triphenylgermanium Chloride
PubChem CID 74197
IUPAC Name chloro(triphenyl)germane
SMILES C1=CC=C(C=C1)[Ge](C2=CC=CC=C2)(C3=CC=CC=C3)Cl

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 339.40
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylchlorogermane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.