accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SBOHHO

CAS RN 162607-15-0

IUPAC Name: (4-methylthiophen-2-yl)boronic acid
Synonyms: 4-methylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 141.98
Molecular Formula: C5H7BO2S
InChi Key: DFUMIZDUIJNUJU-UHFFFAOYSA-N
SMILES: CC1=CSC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results