CAS RN 162635-04-3
IUPAC Name:
(1R,2R,4S)-4-[(2R)-2-[(1R,9S,12S,15R,18R,19R,21R,23S,30S,35R)-1,18-dihydroxy-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-2,3,10,14,20-pentaoxo-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraen-12-yl]propyl]-2-methoxycyclohexyl 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoate
Synonyms:
(1R,2R,4S)-4-[(2R)-2-[(1R,9S,12S,15R,18R,19R,21R,23S,30S,35R)-1,18-dihydroxy-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-2,3,10,14,20-pentaoxo-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraen-12-yl]propyl]-2-methoxycyclohexyl 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoate
Molecular Weight (g/mol):
1030.3
Molecular Formula:
C56H87NO16
InChi Key:
CBPNZQVSJQDFBE-QYCDWSQESA-N
SMILES:
[H]C12CC[C@@H](C)[C@@](O)(O1)C(=O)C(=O)N1CCCC[C@@]1([H])C(=O)O[C@@]([H])(CC(=O)[C@H](C)C=C(C)[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)C=CC=CC=C(C)[C@H](C2)OC)[C@H](C)C[C@@H]1CC[C@@H](OC(=O)C(C)(CO)CO)[C@@H](C1)OC
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Temsirolimus, 99+%
Thermo Scientific Chemicals Temsirolimus, 99+%
CAS: 162635-04-3 Molecular Formula: C56H87NO16,C56H87NO16