accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONHOOH

CAS RN 16284-60-9

IUPAC Name: 2-(3-methylbutanamido)acetic acid
Synonyms: isovalerylglycine
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: ZRQXMKMBBMNNQC-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)NCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results