Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isovalerylglycine 98.0+%, TCI America™
SDP

Catalog No. I03011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03011G 1 g
I030110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03011G Supplier TCI America Supplier No. I03011G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16284-60-9
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00059629
InChI Key ZRQXMKMBBMNNQC-UHFFFAOYSA-N
Synonym N-Isovaleroylglycine
PubChem CID 546304
ChEBI CHEBI:70984
IUPAC Name 2-(3-methylbutanamido)acetic acid
SMILES CC(C)CC(=O)NCC(O)=O

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Isovalerylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.