accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSN

CAS RN 16296-79-0

IUPAC Name: 5-chloro-1-benzothiophene-3-carbonitrile
Synonyms: 5-chloro-1-benzothiophene-3-carbonitrile
Molecular Weight (g/mol): 193.65
Molecular Formula: C9H4ClNS
InChi Key: SXWOPXJSURXXCM-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC=C(C#N)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results