Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chloro-1-benzothiophene-3-carbonitrile, Thermo Scientific™

Catalog No. CC00066DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00066DA 1 g Amber Glass Bottle
CC00066CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00066DA Supplier Thermo Scientific Chemicals Supplier No. CC00066DA
Add to Cart

Chemical Identifiers

CAS 16296-79-0
Molecular Formula C9H4ClNS
Molecular Weight (g/mol) 193.65
MDL Number MFCD02682022
InChI Key SXWOPXJSURXXCM-UHFFFAOYSA-N
Synonym 5-chlorobenzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile,5-chloro, 5-chlorothionaphthene-3-carbonitrile, 5-chloranyl-1-benzothiophene-3-carbonitrile, 5-chloro-1-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 5-chloro
PubChem CID 2776125
IUPAC Name 5-chloro-1-benzothiophene-3-carbonitrile
SMILES ClC1=CC=C2SC=C(C#N)C2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.66
Percent Purity 97%
Chemical Name or Material 5-Chloro-1-benzothiophene-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.