accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 162967-90-0

IUPAC Name: 5,8-dibromo-2,3-bis(4-methoxyphenyl)quinoxaline
Synonyms: 5,8-dibromo-2,3-bis(4-methoxyphenyl)quinoxaline
Molecular Weight (g/mol): 500.19
Molecular Formula: C22H16Br2N2O2
InChi Key: PEHOANSJGUIQOF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(N=C2C(Br)=CC=C(Br)C2=N1)C1=CC=C(OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results