Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,8-Dibromo-2,3-bis(4-methoxyphenyl)quinoxaline 98.0+%, TCI America™
SDP

Catalog No. D4653200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4653200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4653200MG Supplier TCI America Supplier No. D4653200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 162967-90-0
Molecular Formula C22H16Br2N2O2
Molecular Weight (g/mol) 500.19
InChI Key PEHOANSJGUIQOF-UHFFFAOYSA-N
PubChem CID 57712549
IUPAC Name 5,8-dibromo-2,3-bis(4-methoxyphenyl)quinoxaline
SMILES COC1=CC=C(C=C1)C2=NC3=C(C=CC(=C3N=C2C4=CC=C(C=C4)OC)Br)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 500.19
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,8-Dibromo-2,3-bis(4-methoxyphenyl)quinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.