accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163125-34-6

IUPAC Name: 1,1,2,2-tetramethoxycyclohexane
Synonyms: 1,1,2,2-tetramethoxycyclohexane
Molecular Weight (g/mol): 204.27
Molecular Formula: C10H20O4
InChi Key: ZGERRGRQKXRTKD-UHFFFAOYSA-N
SMILES: COC1(CCCCC1(OC)OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results