Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetramethoxycyclohexane, 98%, Thermo Scientific™

Catalog No. AAB2044406
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2044406 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2044406 Supplier Thermo Scientific Chemicals Supplier No. B2044406

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 163125-34-6
Density 1.064
Boiling Point 54°C (0.4mmHg)
Molecular Formula C10H20O4
Quantity 5g
MDL Number MFCD00798587
Synonym cyclohexane,1,1,2,2-tetramethoxy, acmc-1c5qo, 1,2-cyclohexanedione bis dimethyl acetal
InChI Key ZGERRGRQKXRTKD-UHFFFAOYSA-N
SMILES COC1(CCCCC1(OC)OC)OC
IUPAC Name 1,1,2,2-tetramethoxycyclohexane
Molecular Weight (g/mol) 204.266
PubChem CID 2733429
Formula Weight 204.27
Percent Purity 98%
Chemical Name or Material 1,1,2,2-Tetramethoxycyclohexane
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.