accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 1632-76-4

IUPAC Name: 3-methylpyridazine
Synonyms: pyridazine, 3-methyl-
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: MXDRPNGTQDRKQM-UHFFFAOYSA-N
SMILES: CC1=CC=CN=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results