accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHOCH3CH3H3C

CAS RN 16325-63-6

IUPAC Name: 2,4,4-trimethylpentan-1-ol
Synonyms: 2,4,4-trimethylpentan-1-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: ZNRVRWHPZZOTIE-UHFFFAOYNA-N
SMILES: CC(CO)CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results