accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOBrBr

CAS RN 1633-14-3

IUPAC Name: 2,5-dibromocyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-dibromocyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 265.89
Molecular Formula: C6H2Br2O2
InChi Key: KNPAQJBQOIAPBP-UHFFFAOYSA-N
SMILES: BrC1=CC(=O)C(Br)=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results