Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-1,4-benzoquinone 98.0+%, TCI America™
SDP

Catalog No. D22495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22495G 5 g
D22491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22495G Supplier TCI America Supplier No. D22495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1633-14-3
Molecular Formula C6H2Br2O2
Molecular Weight (g/mol) 265.888
MDL Number MFCD00156859
InChI Key KNPAQJBQOIAPBP-UHFFFAOYSA-N
Synonym 2,5-Dibromo-p-quinone
PubChem CID 543093
IUPAC Name 2,5-dibromocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C=C(C1=O)Br)Br

Specifications

Melting Point 193°C
Quantity 5 g
Formula Weight 265.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-1,4-benzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.