accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1633-82-5

IUPAC Name: 3-chloropropane-1-sulfonyl chloride
Synonyms: 3-chloropropane-1-sulfonyl chloride
Molecular Weight (g/mol): 177.04
Molecular Formula: C3H6Cl2O2S
InChi Key: GPKDGVXBXQTHRY-UHFFFAOYSA-N
SMILES: ClCCCS(Cl)(=O)=O
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)