Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Chloropropanesulfonyl chloride

Catalog No. 1251995G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-519-95G 5 g
12-519-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-519-95G Supplier Sigma Aldrich Supplier No. 1251995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-chloropropanesulfonyl chloride was used as starting reagent in the synthesis of the pyrrolidine-based chiral imidazolium ionic liquid
  • It was also used in sulfonation of cross-linked polyethylenimine for selective removal of hg
  • It was used in generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids to yield chiral sultams and sultones

Specifications

CAS 1633-82-5
Density 1.456 g/mL (at 25°C (literature))
Boiling Point 70°C (0.5 mmHg)
Molecular Formula C3H6Cl2O2S
Refractive Index n20/D 1.489 (literature)
Linear Formula Cl(CH2)3 SO2Cl
Quantity 5 g
MDL Number MFCD00007463
Molecular Weight (g/mol) 177.05
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.