accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1634-78-2

H3COOSPOOCH3OCH3OOH3C

CAS RN 1634-78-2

IUPAC Name: 1,4-diethyl 2-[(dimethoxyphosphoryl)sulfanyl]butanedioate
Synonyms: malaoxon liromat malathion-O-analog malaoxone malaoxon (substance) ccris 367 carbethoxy malaoxon maloxon oxycarbophos
Molecular Weight (g/mol): 314.29
Molecular Formula: C10H19O7PS
InChi Key: WSORODGWGUUOBO-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(SP(=O)(OC)OC)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results